accessibility menu, dialog, popup

UserName

CAS RN 628-71-7

H3CCH

CAS RN 628-71-7

IUPAC Name: hept-1-yne
Synonyms: 1-heptyne
Molecular Weight (g/mol): 96.17
Molecular Formula: C7H12
InChi Key: YVXHZKKCZYLQOP-UHFFFAOYSA-N
SMILES: CCCCCC#C
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)