accessibility menu, dialog, popup

UserName

ClN

CAS RN 6280-87-1

IUPAC Name: 5-chloropentanenitrile
Synonyms: 5-chloropentanenitrile
Molecular Weight (g/mol): 117.58
Molecular Formula: C5H8ClN
InChi Key: JSAWFGSXRPCFSW-UHFFFAOYSA-N
SMILES: ClCCCCC#N
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)