accessibility menu, dialog, popup

UserName

FNBrN

CAS RN 62802-38-4

IUPAC Name: 5-bromo-2-fluoropyrimidine
Synonyms: 5-bromo-2-fluoropyrimidine
Molecular Weight (g/mol): 176.98
Molecular Formula: C4H2BrFN2
InChi Key: CTWZYPZCDJKBRS-UHFFFAOYSA-N
SMILES: FC1=NC=C(Br)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results