accessibility menu, dialog, popup

UserName

NNNOHHOOHHCl

CAS RN 6286-29-9

IUPAC Name: 1,3,5-triazinane-1,3,5-triol hydrochloride
Synonyms: sym-triazinetriol hydrochloride
Molecular Weight (g/mol): 171.58
Molecular Formula: C3H10ClN3O3
InChi Key: VRHJXINSVWQXEB-UHFFFAOYSA-N
SMILES: Cl.ON1CN(O)CN(O)C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results