accessibility menu, dialog, popup

UserName

H3COOH2NOOCH3

CAS RN 6296-99-7

IUPAC Name: 1,3-diethyl 2-(aminomethylidene)propanedioate
Synonyms: 1,3-diethyl 2-(aminomethylidene)propanedioate
Molecular Weight (g/mol): 187.2
Molecular Formula: C8H13NO4
InChi Key: WDTWMEZMHUEZKH-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=CN)C(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results