CAS RN 63038-27-7
IUPAC Name:
methyl 2-amino-3,3-dimethylbutanoate hydrochloride
Synonyms:
methyl 2-amino-3,3-dimethylbutanoate hydrochloride
Molecular Weight (g/mol):
181.66
Molecular Formula:
C7H16ClNO2
InChi Key:
HRTQWUHFSXVRPY-UHFFFAOYNA-N
SMILES:
Cl.COC(=O)C(N)C(C)(C)C
1
–
4
of
4
results
2
L-tert-Leucine Methyl Ester Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
L-tert-Leucine Methyl Ester Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
L-tert-Leucine Methyl Ester Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.