accessibility menu, dialog, popup

UserName

CAS RN 631-67-4

H3CNCH3CH3S

CAS RN 631-67-4

IUPAC Name: N,N-dimethylethanethioamide
Synonyms: ethanethioamide, N,N-dimethyl-
Molecular Weight (g/mol): 103.18
Molecular Formula: C4H9NS
InChi Key: LKNQXZAHNDFIQY-UHFFFAOYSA-N
SMILES: CN(C)C(C)=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results