accessibility menu, dialog, popup

UserName

H3CBOHHO

CAS RN 63139-21-9

IUPAC Name: (4-ethylphenyl)boronic acid
Synonyms: 4-ethylphenylboronic acid
Molecular Weight (g/mol): 149.98
Molecular Formula: C8H11BO2
InChi Key: RZCPLOMUUCFPQA-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)B(O)O
Percent Purity 
  • (6)
  • (7)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (3)
  • (7)
  • (6)
  • (7)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)