accessibility menu, dialog, popup

UserName

IBr

CAS RN 63279-58-3

IUPAC Name: 1-bromo-4-iodonaphthalene
Synonyms: 1-bromo-4-iodonaphthalene
Molecular Weight (g/mol): 332.97
Molecular Formula: C10H6BrI
InChi Key: HQHHKYXPFKHLBF-UHFFFAOYSA-N
SMILES: BrC1=CC=C(I)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results