accessibility menu, dialog, popup

UserName

NH2NOON

CAS RN 6393-40-4

IUPAC Name: 4-amino-3-nitrobenzonitrile
Synonyms: 4-amino-3-nitrobenzonitrile
Molecular Weight (g/mol): 163.14
Molecular Formula: C7H5N3O2
InChi Key: JAHADAZIDZMHOP-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1[N+]([O-])=O)C#N
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)