accessibility menu, dialog, popup

UserName

HOOS

CAS RN 6435-75-2

IUPAC Name: 4,5,6,7-tetrahydro-2-benzothiophene-1-carboxylic acid
Synonyms: 4,5,6,7-tetrahydro-2-benzothiophene-1-carboxylic acid
Molecular Weight (g/mol): 182.24
Molecular Formula: C9H10O2S
InChi Key: ZBNTUDLPJCXPNF-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2CCCCC2=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results