accessibility menu, dialog, popup

UserName

CH3OOOH

CAS RN 64354-50-3

IUPAC Name: 5-methyl-2-phenylfuran-3-carboxylic acid
Synonyms: 5-methyl-2-phenylfuran-3-carboxylic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C12H10O3
InChi Key: CASZQQKUMVLKBY-UHFFFAOYSA-N
SMILES: CC1=CC(C(O)=O)=C(O1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results