accessibility menu, dialog, popup

UserName

H3CCH3OHCH3OO

CAS RN 64366-70-7

IUPAC Name: 2-ethylhexan-1-ol; 2-methyloxirane; oxirane
Synonyms: 2-ethylhexanol; ethylene oxide; propylene oxide
Molecular Weight (g/mol): 232.36
Molecular Formula: C13H28O3
InChi Key: LLCXFSRLSYMSPW-UHFFFAOYNA-N
SMILES: C1CO1.CC1CO1.CCCCC(CC)CO
Quantity 
  • (1)
  • (1)
  • (1)
pH 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results