CAS RN 64502-89-2
CAS RN 64502-89-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-hydroxypentan-2-one, TRC
CAS: 64502-89-2 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 IUPAC Name: 1-hydroxypentan-2-one SMILES: CCCC(=O)CO