accessibility menu, dialog, popup

UserName

H3COOOONClClCl

CAS RN 64700-56-7

IUPAC Name: 2-butoxyethyl 2-[(3,5,6-trichloropyridin-2-yl)oxy]acetate
Synonyms: triclopyr ester triclopyr, butoxyethyl ester
Molecular Weight (g/mol): 356.62
Molecular Formula: C13H16Cl3NO4
InChi Key: IVDRCZNHVGQBHZ-UHFFFAOYSA-N
SMILES: CCCCOCCOC(=O)COC1=NC(Cl)=C(Cl)C=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results