accessibility menu, dialog, popup

UserName

HCHCVBrBrBr

CAS RN 64815-29-8

IUPAC Name: bis(cyclopenta-2,4-dien-1-ide) vanadium tribromide
Synonyms: bis(cyclopentadienide) vanadium tribromide
Molecular Weight (g/mol): 420.85
Molecular Formula: C10H10Br3V
InChi Key: QHYHAEAMZXFABY-UHFFFAOYSA-K
SMILES: [V].[Br-].[Br-].[Br-].[CH-]1C=CC=C1.[CH-]1C=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results