accessibility menu, dialog, popup

UserName

ONHO

CAS RN 64943-90-4

IUPAC Name: 6-phenyl-2H,5H-[1,3]dioxolo[4,5-f]indole
Synonyms: 6-phenyl-2H,5H-[1,3]dioxolo[4,5-f]indole
Molecular Weight (g/mol): 237.26
Molecular Formula: C15H11NO2
InChi Key: OURPDRQDIRKULF-UHFFFAOYSA-N
SMILES: C1OC2=C(O1)C=C1C=C(NC1=C2)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results