accessibility menu, dialog, popup

UserName

OONN

CAS RN 64951-08-2

IUPAC Name: imidazo[1,2-a]pyridine-2-carboxylate
Synonyms: imidazo[1,2-a]pyridine-2-carboxylate
Molecular Weight (g/mol): 161.14
Molecular Formula: C8H5N2O2
InChi Key: WQLJLPDGSLZYEP-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CN2C=CC=CC2=N1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)