accessibility menu, dialog, popup

UserName

CAS RN 65264-05-3

IUPAC Name: 4-ethyl-N'-hydroxybenzene-1-carboximidamide
Synonyms: 4-ethyl-N'-hydroxybenzenecarboximidamide
Molecular Weight (g/mol): 164.21
Molecular Formula: C9H12N2O
InChi Key: RSGJHKJTRAXLPF-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)C(N)=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results