accessibility menu, dialog, popup

UserName

OON

CAS RN 6537-46-8

IUPAC Name: 10H-chromeno[3,2-b]pyridin-10-one
Synonyms: chromeno[3,2-b]pyridin-10-one
Molecular Weight (g/mol): 197.19
Molecular Formula: C12H7NO2
InChi Key: YAUODDQCNOBEDJ-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2OC2=CC=CN=C12
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (2)