accessibility menu, dialog, popup

UserName

H3COOSNH3C

CAS RN 656-53-1

IUPAC Name: 2-(4-methyl-1,3-thiazol-5-yl)ethyl acetate
Synonyms: 2-(4-methyl-1,3-thiazol-5-yl)ethyl acetate
Molecular Weight (g/mol): 185.24
Molecular Formula: C8H11NO2S
InChi Key: CRTCWNPLKVVXIX-UHFFFAOYSA-N
SMILES: CC(=O)OCCC1=C(C)N=CS1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results