accessibility menu, dialog, popup

UserName

CAS RN 65706-99-2

OOOOHNHOO

CAS RN 65706-99-2

IUPAC Name: 5-(benzyloxy)-4-{[(benzyloxy)carbonyl]amino}-5-oxopentanoic acid
Synonyms: 5-(benzyloxy)-4-{[(benzyloxy)carbonyl]amino}-5-oxopentanoic acid
Molecular Weight (g/mol): 371.39
Molecular Formula: C20H21NO6
InChi Key: VWHKODOUMSMUAF-UHFFFAOYNA-N
SMILES: OC(=O)CCC(NC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)