accessibility menu, dialog, popup

UserName

ClClNCl

CAS RN 6575-05-9

IUPAC Name: 2,4,6-trichlorobenzonitrile
Synonyms: 2,4,6-trichlorobenzonitrile
Molecular Weight (g/mol): 206.45
Molecular Formula: C7H2Cl3N
InChi Key: PGODHCIOIPODFE-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(C#N)C(Cl)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)