accessibility menu, dialog, popup

UserName

ClNCSCl

CAS RN 6590-96-1

IUPAC Name: 2,4-dichloro-1-isothiocyanatobenzene
Synonyms: 2,4-dichloro-1-isothiocyanatobenzene
Molecular Weight (g/mol): 204.07
Molecular Formula: C7H3Cl2NS
InChi Key: WVBNZZHGECFCSH-UHFFFAOYSA-N
SMILES: ClC1=CC=C(N=C=S)C(Cl)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results