accessibility menu, dialog, popup

UserName

HClCH3OHNH2N

CAS RN 66-83-1

IUPAC Name: hydrogen 2-(5-methoxy-1H-indol-3-yl)ethan-1-amine chloride
Synonyms: hydrogen 5 methoxytryptamine chloride
Molecular Weight (g/mol): 226.7
Molecular Formula: C11H15ClN2O
InChi Key: TXVAYRSEKRMEIF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC=C2NC=C(CCN)C2=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)