accessibility menu, dialog, popup

UserName

CAS RN 6628-83-7

H2NO

CAS RN 6628-83-7

IUPAC Name: 1-(oxan-2-yl)methanamine
Synonyms: 1-(oxan-2-yl)methanamine
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: NYGCBQKDTGBHSC-UHFFFAOYNA-N
SMILES: NCC1CCCCO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results