accessibility menu, dialog, popup

UserName

CAS RN 6638-05-7

CH3OCH3OCH3HO

CAS RN 6638-05-7

IUPAC Name: 2,6-dimethoxy-4-methylphenol
Synonyms: 2,6-dimethoxy-4-methylphenol
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: ZFBNNSOJNZBLLS-UHFFFAOYSA-N
SMILES: COC1=CC(C)=CC(OC)=C1O
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)