accessibility menu, dialog, popup

UserName

CH3H3CH3COHO(E)

CAS RN 66634-97-7

IUPAC Name: (2E)-2,4-dimethylpent-2-enoic acid
Synonyms: (2E)-2,4-dimethylpent-2-enoic acid
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: DMHLGGQHOSTMJG-GQCTYLIASA-N
SMILES: CC(C)\C=C(/C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results