CAS RN 668270-11-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-Linagliptin, TRC
CAS: 668270-11-9 Molecular Formula: C25H28N8O2 Molecular Weight (g/mol): 472.54 Synonym: 8-[(3S)-3-Amino-1-piperidinyl]-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-1H-purine-2,6-dione IUPAC Name: 8-[(3S)-3-aminopiperidin-1-yl]-7-but-2-ynyl-3-methyl-1-[(4-methylquinazolin-2-yl)methyl]purine-2,6-dione SMILES: O=C(N(C)C(N=C(N1C[C@@H](N)CCC1)N2CC#CC)=C2C3=O)N3CC4=NC5=CC=CC=C5C(C)=N4