CAS RN 6683-92-7
CAS RN 6683-92-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Phenyl-2-pentanone, TRC
CAS: 6683-92-7 Molecular Formula: C11 H14 O Molecular Weight (g/mol): 162.23 Synonym: 1-Phenyl-2-pentanone,Benzyl propyl ketone IUPAC Name: 1-phenylpentan-2-one SMILES: CCCC(=O)Cc1ccccc1