accessibility menu, dialog, popup

UserName

OOBr

CAS RN 6698-13-1

IUPAC Name: 4-bromo-2H-1,3-benzodioxole
Synonyms: 4-bromo-2H-1,3-benzodioxole
Molecular Weight (g/mol): 201.02
Molecular Formula: C7H5BrO2
InChi Key: VZPMQHSDFWAZHP-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=C1OCO2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results