accessibility menu, dialog, popup

UserName

H3COOH

CAS RN 66990-32-7

IUPAC Name: pentacosa-10,12-diynoic acid
Synonyms: pentacosa-10,12-diynoic acid
Molecular Weight (g/mol): 374.61
Molecular Formula: C25H42O2
InChi Key: ZPUDRBWHCWYMQS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC#CC#CCCCCCCCCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results