accessibility menu, dialog, popup

UserName

H3CPCH3

CAS RN 672-66-2

IUPAC Name: dimethyl(phenyl)phosphane
Synonyms: dimethylphenylphosphine (dimethyl)-phenylphosphine Me2PPh
Molecular Weight (g/mol): 138.15
Molecular Formula: C8H11P
InChi Key: HASCQPSFPAKVEK-UHFFFAOYSA-N
SMILES: CP(C)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)