accessibility menu, dialog, popup

UserName

CAS RN 675-10-5

CH3OHOO

CAS RN 675-10-5

IUPAC Name: 2-hydroxy-6-methyl-4H-pyran-4-one
Synonyms: triacetic acid lactone
Molecular Weight (g/mol): 126.11
Molecular Formula: C6H6O3
InChi Key: OOKCZXGEYPSNIM-UHFFFAOYSA-N
SMILES: CC1=CC(=O)C=C(O)O1
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)