accessibility menu, dialog, popup

UserName

CAS RN 67531-68-4

IUPAC Name: ethyl 3-isocyanatobenzoate
Synonyms: ethyl 3-isocyanatobenzoate
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: UPXGGIRFDZENRD-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(=CC=C1)N=C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results