CAS RN 675602-91-2
IUPAC Name:
2,3,5-trifluoropyridine-4-carboxylic acid
Synonyms:
2,3,5-trifluoropyridine-4-carboxylic acid
Molecular Weight (g/mol):
177.08
Molecular Formula:
C6H2F3NO2
InChi Key:
UKLNELOJJKZMLJ-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(F)C(F)=NC=C1F
1
–
1
of
1
results
1
2,3,5-Trifluoropyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals
CAS: 675602-91-2 Molecular Formula: C6H2F3NO2 Molecular Weight (g/mol): 177.082 MDL Number: MFCD03840205 InChI Key: UKLNELOJJKZMLJ-UHFFFAOYSA-N Synonym: 2,3,5-trifluoroisonicotinic acid,4-pyridinecarboxylic acid, 2,3,5-trifluoro,2,3,5-trifluoropyridine-4-carboxylic acid PubChem CID: 2782836 IUPAC Name: 2,3,5-trifluoropyridine-4-carboxylic acid SMILES: C1=C(C(=C(C(=N1)F)F)C(=O)O)F