accessibility menu, dialog, popup

UserName

H3COONNBr

CAS RN 67625-37-0

IUPAC Name: ethyl 6-bromoimidazo[1,2-a]pyridine-2-carboxylate
Synonyms: ethyl 6-bromoimidazo[1,2-a]pyridine-2-carboxylate
Molecular Weight (g/mol): 269.1
Molecular Formula: C10H9BrN2O2
InChi Key: MWDCKDQQAFZDOH-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN2C=C(Br)C=CC2=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results