accessibility menu, dialog, popup

UserName

CAS RN 6775-01-5

IUPAC Name: bis(3-azaniumylpropyl)(phenyl)phosphane
Synonyms: bis(3-ammoniopropyl)(phenyl)phosphane
Molecular Weight (g/mol): 226.3
Molecular Formula: C12H23N2P
InChi Key: OEHBMEXOPYWPKQ-UHFFFAOYSA-P
SMILES: [NH3+]CCCP(CCC[NH3+])C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results