accessibility menu, dialog, popup

UserName

CH3H3CCl

CAS RN 6781-98-2

IUPAC Name: 2-chloro-1,3-dimethylbenzene
Synonyms: 2-chloro-1,3-dimethylbenzene
Molecular Weight (g/mol): 140.61
Molecular Formula: C8H9Cl
InChi Key: VDXLAYAQGYCQEO-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)