accessibility menu, dialog, popup

UserName

CH3SnH3CCH3CH2H3C

CAS RN 67883-62-9

IUPAC Name: tributyl(2-methylprop-2-en-1-yl)stannane
Synonyms: tributyl(2-methylprop-2-en-1-yl)stannane
Molecular Weight (g/mol): 345.16
Molecular Formula: C16H34Sn
InChi Key: CYIUFVYZGOCGEV-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)CC(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results