accessibility menu, dialog, popup

UserName

CAS RN 68083-14-7

IUPAC Name: 3,3-dimethyl-5,5-diphenyl-2,4-dioxa-3,5-disilahexane
Synonyms: 3,3-dimethyl-5,5-diphenyl-2,4-dioxa-3,5-disilahexane
Molecular Weight (g/mol): 302.52
Molecular Formula: C16H22O2Si2
InChi Key: ARWRSWALIGRKQA-UHFFFAOYSA-N
SMILES: CO[Si](C)(C)O[Si](C)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results