accessibility menu, dialog, popup

UserName

CH3NHNO

CAS RN 68282-49-5

IUPAC Name: 2-butyl-1H-imidazole-5-carbaldehyde
Synonyms: 2-butyl-3H-imidazole-4-carbaldehyde
Molecular Weight (g/mol): 152.2
Molecular Formula: C8H12N2O
InChi Key: PTHGVOCFAZSNNA-UHFFFAOYSA-N
SMILES: CCCCC1=NC=C(N1)C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results