accessibility menu, dialog, popup

UserName

CAS RN 683-60-3

IUPAC Name: sodium propan-2-olate
Synonyms: sodium propan-2-olate
Molecular Weight (g/mol): 82.08
Molecular Formula: C3H7NaO
InChi Key: WBQTXTBONIWRGK-UHFFFAOYSA-N
SMILES: [Na+].CC(C)[O-]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results