accessibility menu, dialog, popup

UserName

OH2NHCl

CAS RN 683233-12-7

IUPAC Name: 1-(oxan-2-yl)methanamine hydrochloride
Synonyms: 1-(oxan-2-yl)methanamine hydrochloride
Molecular Weight (g/mol): 151.63
Molecular Formula: C6H14ClNO
InChi Key: NGHRATGQJXKQIQ-UHFFFAOYNA-N
SMILES: Cl.NCC1CCCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results