accessibility menu, dialog, popup

UserName

ON

CAS RN 6837-24-7

IUPAC Name: 1-cyclohexylpyrrolidin-2-one
Synonyms: N-cyclohexyl-2-pyrrolidone
Molecular Weight (g/mol): 167.25
Molecular Formula: C10H17NO
InChi Key: PZYDAVFRVJXFHS-UHFFFAOYSA-N
SMILES: O=C1CCCN1C1CCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results