accessibility menu, dialog, popup

UserName

OHOHOFFF

CAS RN 684-07-1

IUPAC Name: 3,3,3-trifluoro-2-hydroxypropanoic acid
Synonyms: 3,3,3-trifluoro-2-hydroxypropanoic acid
Molecular Weight (g/mol): 144.05
Molecular Formula: C3H3F3O3
InChi Key: BVKGUTLIPHZYCX-UHFFFAOYNA-N
SMILES: OC(C(O)=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)