accessibility menu, dialog, popup

UserName

H3CNHOHNO

CAS RN 6843-49-8

IUPAC Name: 5-methyl-5-phenylimidazolidine-2,4-dione
Synonyms: 5-methyl-5-phenylhydantoin
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: JNGWGQUYLVSFND-UHFFFAOYNA-N
SMILES: CC1(NC(=O)NC1=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)