accessibility menu, dialog, popup

UserName

CH3NH3CH3CH3CH2OCl

CAS RN 68696-18-4

IUPAC Name: tetraethylazanium hydrate chloride
Synonyms: tetraethylammonium hydrate chloride
Molecular Weight (g/mol): 183.72
Molecular Formula: C8H22ClNO
InChi Key: CALLTGJPWMIDPC-UHFFFAOYSA-M
SMILES: O.[Cl-].CC[N+](CC)(CC)CC
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)