accessibility menu, dialog, popup

UserName

CAS RN 68797-61-5

ClNNClBr

CAS RN 68797-61-5

IUPAC Name: 5-bromo-4,6-dichloropyrimidine
Synonyms: 5-bromo-4,6-dichloropyrimidine
Molecular Weight (g/mol): 227.87
Molecular Formula: C4HBrCl2N2
InChi Key: BKLHHTTZTSYHBV-UHFFFAOYSA-N
SMILES: ClC1=NC=NC(Cl)=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results