accessibility menu, dialog, popup

UserName

CAS RN 68832-13-3

IUPAC Name: (2R)-2-(hydroxymethyl)pyrrolidin-1-ium
Synonyms: prolinol
Molecular Weight (g/mol): 102.16
Molecular Formula: C5H12NO
InChi Key: HVVNJUAVDAZWCB-RXMQYKEDSA-O
SMILES: OC[C@H]1CCC[NH2+]1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)